Ethyl-2-(2-formylphenoxy)butanoate ≥95%
Supplier: Matrix Scientific
044874-5G
044874-1G
044874-500MG
101867-992EA
532.31
USD
101867-992
101867-990
101867-988
Ethyl-2-(2-formylphenoxy)butanoate ≥95%
Ethyl-2-(2-formylphenoxy)butanoate
Hazard: Irritant
Formula:
C₁₃H₁₆O₄ Boiling Pt: 104…106 °C (0.05 mmHg) |
MDL Number:
MFCD03425787 CAS Number: 86602-60-0 |
Specification Test Results
Boiling point | 104-106°C/0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...