Pentafluorophenyl 1-ethylenesulfonate ≥95%

Supplier: Matrix Scientific

044654-5G 044654-1G 044654-500MG
101866-936EA 802.7 USD
101866-936 101866-932 101866-934
Pentafluorophenyl 1-ethylenesulfonate ≥95%
Pentafluorophenyl 1-ethylenesulphonate

Hazard: Irritant/Refrigerate

Order Now

Specification Test Results

Boiling point 74°C/.06mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR