Methyl-2-(1H-1,2,4-triazol-1-yl)acetate ≥95%

Supplier: Matrix Scientific

044562-500MG 044562-5G 044562-1G
101866-484EA 350.77 USD
101866-484 101866-486 101866-482
Methyl-2-(1H-1,2,4-triazol-1-yl)acetate ≥95%
Methyl-2-(1H-1,2,4-triazol-1-yl)acetate

Hazard: Irritant

Formula: C₅H₇N₃O₂
MW: 141.13 g/mol
Boiling Pt: 80 °C (0.3 mmHg)
MDL Number: MFCD02186570
CAS Number: 106535-16-4

Order Now

Specification Test Results

Boiling point 80°C/0.3mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR