tert-Butyl-4-fluorobenzoate ≥96%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 4-fluorobenzoate
001730-25G
001730-5G
101782-426EA
41.54
USD
101782-426
101782-428
tert-Butyl-4-fluorobenzoate ≥96%
tert-Butyl 4-fluorobenzoate
Hazard: Irritant
Formula:
C₁₁H₁₃FO₂ MW: 196.2181 g/mol Boiling Pt: 51…52 °C (0.4 mmHg) |
MDL Number:
MFCD01320759 CAS Number: 58656-98-7 |
Specification Test Results
Boiling point | 51-52°C/0.4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...