4-(Trifluormethyl)nicotinonitrile ≥95%

Supplier: Matrix Scientific

001381-1G 001381-5G
101781-534EA 320.85 USD
101781-534 101781-532
4-(Trifluormethyl)nicotinonitrile ≥95%
4-(Trifluormethyl)nicotinonitrile

Hazard: Irritant

Formula: C₇H₃F₃N₂
Boiling Pt: 75…77 °C (15 mmHg)
MDL Number: MFCD00153107
CAS Number: 13600-43-6

Order Now

Specification Test Results

Boiling point 75-77°C/15m

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR