3-Nitrophenylsulfur pentafluoride ≥97%
Supplier: Matrix Scientific
003249-1G
101785-456EA
23.08
USD
101785-456
3-Nitrophenylsulfur pentafluoride ≥97%
3-Nitrophenylsulphur pentafluoride
Hazard: Toxic/Irritant
Formula:
C₆H₄F₅NO₂S Boiling Pt: 106 °C (2 mmHg) Density: 1.662 |
MDL Number:
MFCD00221621 CAS Number: 2613-26-5 UN: 2810 ADR: 6.1,III |
Specification Test Results
Boiling point | 106°C/2mm |
ND20 | 1.476 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...