4-Chlorobenzyl thiocyanate ≥98%
Supplier: Matrix Scientific
056949-100G
056949-25G
056949-5G
101911-836EA
87.69
USD
101911-836
101911-838
101911-840
4-Chlorobenzyl thiocyanate ≥98%
4-Chlorobenzyl thiocyanate
Hazard: Irritant-Harmful
Formula:
C₈H₆ClNS Boiling Pt: 108 °C (1 mmHg) Density: 1.26 |
MDL Number:
MFCD01680436 CAS Number: 2082-64-6 |
Specification Test Results
Boiling point | 108°C/1mm |
Density | 1.26 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...