1-(m-Tolyl)imidazole ≥98%
Supplier: Matrix Scientific
056828-1G
101911-484EA
141.54
USD
101911-484
1-(m-Tolyl)imidazole ≥98%
1-(m-Tolyl)imidazole
Hazard: Irritant
Formula:
C₁₀H₁₀N₂ Boiling Pt: 145 °C (1 mmHg) Density: 1.1 |
CAS Number:
25364-43-6 |
Specification Test Results
Boiling point | 145°C/1mm |
Density | 1.1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...