3-(Pentylamino)propionitrile ≥98%
056792-10G
101911-410EA
1070.08
USD
101911-410
3-(Pentylamino)propionitrile ≥98%
3-(Pentylamino)propionitrile
Hazard: Irritant
Specification Test Results
Boiling point | 129°C/20mm |
Density | 0.87 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...