2,3-Dimethoxybenzyl isothiocyanate ≥98%

Supplier: Matrix Scientific

056710-5G
101911-246EA 206.15 USD
101911-246
2,3-Dimethoxybenzyl isothiocyanate ≥98%
2,3-Dimethoxybenzyl isothiocyanate

Hazard: Irritant-Harmful

Formula: C₁₀H₁₁NO₂S
Boiling Pt: 127 °C (5 mmHg)
Density: 1.2
MDL Number: MFCD00060402
CAS Number: 34964-55-1

Order Now

Specification Test Results

Boiling point 127°C/5mm
Density 1.2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR