Crotonyl isothiocyanate

Supplier: Matrix Scientific

056692-5G
101911-214EA 273.85 USD
101911-214
Crotonyl isothiocyanate
Crotonyl isothiocyanate

Hazard: Irritant-Harmful

Formula: C₅H₅NOS
Boiling Pt: 78…80 °C (1 mmHg)
Density: 1.1
MDL Number: MFCD08461770
CAS Number: 60034-28-8

Order Now

Specification Test Results

Boiling point 78-80°C/1mm
Density 1.1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR