3-(4-Morpholino)propyl isothiocyanate ≥98%

Supplier: Matrix Scientific

056608-5G
101911-030EA 121.54 USD
101911-030
3-(4-Morpholino)propyl isothiocyanate ≥98%
3-(4-Morpholino)propyl isothiocyanate

Hazard: Irritant-Harmful

Formula: C₈H₁₄N₂OS
Boiling Pt: 183 °C (13 mmHg)
Density: 1.08
MDL Number: MFCD00041222
CAS Number: 32813-50-6

Order Now

Specification Test Results

Boiling point 183°C/13mm
Density 1.08

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR