2-(4-Morpholino)ethyl isothiocyanate ≥98%

Supplier: Matrix Scientific

056607-5G
101911-028EA 103.5 USD
101911-028
2-(4-Morpholino)ethyl isothiocyanate ≥98%
2-(4-Morpholino)ethyl isothiocyanate

Hazard: Irritant-Harmful

Formula: C₇H₁₂N₂OS
Boiling Pt: 163 °C (10 mmHg)
Density: 1.11
MDL Number: MFCD00041221
CAS Number: 63224-35-1

Order Now

Specification Test Results

Boiling point 163°C/10mm
Density 1.11

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR