4-Methylphenethyl isothiocyanate ≥99%

Supplier: Matrix Scientific

056606-1G
101911-026EA 81.54 USD
101911-026
4-Methylphenethyl isothiocyanate ≥99%
4-Methylphenethyl isothiocyanate

Hazard: Irritant-Harmful

Formula: C₁₀H₁₁NS
Boiling Pt: 155 °C (20 mmHg)
Density: 1.07
MDL Number: MFCD00046823
CAS Number: 13203-39-9

Order Now

Specification Test Results

Boiling point 155°C/20mm
Density 1.07

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR