3-Methylbutyl isothiocyanate ≥98%

Supplier: Matrix Scientific

056598-1G 056598-5G
101910-998EA 166.75 USD
101910-998 101910-996
3-Methylbutyl isothiocyanate ≥98%
3-Methylbutyl isothiocyanate

Hazard: Irritant-Harmful

Formula: C₆H₁₁NS
Boiling Pt: 183…184 °C
Density: 0.94
MDL Number: MFCD00060419
CAS Number: 628-03-5

Order Now

Specification Test Results

Boiling point 183-184°C
Density 0.94

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR