4-Methylbenzyl isothiocyanate ≥99%

Supplier: Matrix Scientific

056597-5G
101910-994EA 85.1 USD
101910-994
4-Methylbenzyl isothiocyanate ≥99%
4-Methylbenzyl isothiocyanate

Hazard: Irritant-Harmful

Formula: C₉H₉NS
Boiling Pt: 97 °C (1 mmHg)
Density: 1.08
MDL Number: MFCD00041118
CAS Number: 3694-46-0

Order Now

Specification Test Results

Boiling point 97°C/1mm
Density 1.08

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR