2-Methylbenzyl isothiocyanate ≥99%

Supplier: Matrix Scientific

056595-1G 056595-5G
101910-988EA 312.31 USD
101910-988 101910-986
2-Methylbenzyl isothiocyanate ≥99%
2-Methylbenzyl isothiocyanate

Hazard: Irritant-Harmful

Formula: C₉H₉NS
Boiling Pt: 136 °C (2 mmHg)
Density: 1.09
MDL Number: MFCD00060404
CAS Number: 16735-69-6

Order Now

Specification Test Results

Boiling point 136°C/2mm
Density 1.09

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR