2-Isopropyl-6-methylphenyl isothiocyanate ≥95%

Supplier: Matrix Scientific

056569-1G 056569-5G
101910-904EA 38.46 USD
101910-904 101910-906
2-Isopropyl-6-methylphenyl isothiocyanate ≥95%
2-Isopropyl-6-methylphenyl isothiocyanate

Hazard: Irritant-Harmful

Formula: C₁₁H₁₃NO
MW: 175.23 g/mol
Boiling Pt: 92 °C (3 mmHg)
Density: 1.02
MDL Number: MFCD00060370
CAS Number: 306935-86-4

Order Now

Specification Test Results

Boiling point 92°C/3mm
Density 1.02

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR