2-Heptyl isothiocyanate ≥99%

Supplier: Matrix Scientific

056566-5G
101910-896EA 150.77 USD
101910-896
2-Heptyl isothiocyanate ≥99%
2-Heptyl isothiocyanate

Hazard: Irritant-Harmful

Formula: C₈H₁₅NS
Boiling Pt: 100 °C (10 mmHg)
Density: 0.91
MDL Number: MFCD00060399
CAS Number: 21663-51-4

Order Now

Specification Test Results

Boiling point 100°C/10mm
Density 0.91

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR