Fluoromethyl-1,1,1,3,3,3-hexafluoroisopropyl ether ≥99%

Supplier: Matrix Scientific

007183-25G 007183-5G
101792-146EA 241.54 USD
101792-146 101792-144
Fluoromethyl-1,1,1,3,3,3-hexafluoroisopropyl ether ≥99%
Fluoromethyl-1,1,1,3,3,3-hexafluoroisopropyl ether

Purity: 99%
CAS: 28523-86-6
MDL Number: MFCD00153189
Molecular Formula: C4H3F7O
Molecular Weight: 200,06
Boiling Point: 58°C
Hazard: Irritant/Moisture Sensitive/Stored under Argon

Formula: C₄H₃F₇O
MW: 200.06 g/mol
Boiling Pt: 58 °C
Density: 1.505
MDL Number: MFCD00153189
CAS Number: 28523-86-6

Order Now

Specification Test Results

Boiling point 58°C
Density 1.505

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR