4-Isopropylphenylacetonitrile ≥96%

Supplier: Matrix Scientific

Synonyms: 2-(4-Isopropylphenyl)acetonitrile

006651-5G 006651-25G
101790-866EA 176.92 USD
101790-866 101790-868
4-Isopropylphenylacetonitrile ≥96%
4-Isopropylphenylacetonitrile

Hazard: Irritant/Irritant-Harmful

Formula: C₁₁H₁₃N
MW: 159.23 g/mol
Boiling Pt: 150 °C (25 mmHg)
Density: 0.96
MDL Number: MFCD00040892
CAS Number: 4395-87-3
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 150°C/25mm
Density 0.96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR