4-Phenoxyphenethylamine ≥98%
Supplier: Matrix Scientific
005693-5G
005693-1G
101789-364EA
536.92
USD
101789-364
101789-362
4-Phenoxyphenethylamine ≥98%
4-Phenoxyphenethylamine
Hazard: Irritant/Irritant-Harmful/Stored under Argon
Specification Test Results
Boiling point | 125°C/1mm |
Density | 1.09 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...