3-(4-Bromophenyl)propiononitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 3-(4-Bromophenyl)propanenitrile

004943-5G
101788-078EA 345 USD
101788-078
3-(4-Bromophenyl)propiononitrile ≥98%
3-(4-Bromophenyl)propiononitrile

Hazard: Irritant/Irritant-Harmful

Formula: C₉H₈BrN
MW: 210.0705 g/mol
Boiling Pt: 118 °C (1 mmHg)
Density: 1.42
MDL Number: MFCD01310795
CAS Number: 57775-08-3

Order Now

Specification Test Results

Boiling point 118°C/1mm
Density 1.42

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR