2,3-Dibromo-1,1,1,4,4,4-hexafluorobutane ≥97%

Supplier: Matrix Scientific

004525-500MG
101787-384EA 118.46 USD
101787-384
2,3-Dibromo-1,1,1,4,4,4-hexafluorobutane ≥97%
2,3-Dibromo-1,1,1,4,4,4-hexafluorobutane

Hazard: Irritant/Light Sensitive

Order Now

Specification Test Results

Boiling point 62°C/300mm
Density 1.8

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR