3-(Hexylamino)propionitrile ≥99%
Supplier: Matrix Scientific
Synonyms:
3-(Hexylamino)propanenitrile
004511-5G
101787-358EA
38.46
USD
101787-358
3-(Hexylamino)propionitrile ≥99%
3-(Hexylamino)propionitrile
Hazard: Irritant-Harmful
Specification Test Results
Boiling point | 130-134°C/1 |
Density | 0.86 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...