3-(Butylamino)propionitrile ≥99%
Supplier: Matrix Scientific
004510-25G
101787-356EA
176.92
USD
101787-356
3-(Butylamino)propionitrile ≥99%
3-(Butylamino)propionitrile
Hazard: Irritant/Irritant-Harmful
Specification Test Results
Boiling point | 104-106°C/1 |
Density | 0.86 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...