Butyl fluoroacetate ≥98%

Supplier: Matrix Scientific

Synonyms: Fluoroacetic Acid Butyl Ester

004379-25G
101787-128EA 536.92 USD
101787-128
Butyl fluoroacetate ≥98%
Butyl fluoroacetate

Purity: 98%
CAS: 371-49-3
MDL Number: MFCD00042194
Molecular Formula: C6H11FO2
Molecular Weight: 134,15
Boiling Point: 154°C
Hazard: Irritant

Formula: C₆H₁₁FO₂
MW: 134.15 g/mol
Boiling Pt: 154 °C
Density: 1.02
MDL Number: MFCD00042194
CAS Number: 371-49-3

Order Now

Specification Test Results

Boiling point 154°C
Density 1.02

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR