2-Chlorodiphenylmethane ≥98%
Supplier: Matrix Scientific
004226-1G
004226-5G
101787-002EA
560
USD
101787-002
101787-000
2-Chlorodiphenylmethane ≥98%
2-Chlorodiphenylmethane
Hazard: Irritant
Specification Test Results
Boiling point | 97°C/1mm |
Density | 1.12 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...