2-Fluorodiphenylmethane ≥98%

Supplier: Matrix Scientific

004225-1G 004225-5G
101786-996EA 43.7 USD
101786-996 101786-998
2-Fluorodiphenylmethane ≥98%
2-Fluorodiphenylmethane

Hazard: Irritant

Formula: C₁₃H₁₁F
Boiling Pt: 76 °C (1 mmHg)
Density: 1.08
MDL Number: MFCD00079758
CAS Number: 3794-15-8

Order Now

Specification Test Results

Boiling point 76°C/1mm
Density 1.08

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR