3-Ethoxyphenylacetonitrile ≥99%
Supplier: Matrix Scientific
003898-1G
003898-5G
101786-394EA
213.85
USD
101786-394
101786-396
3-Ethoxyphenylacetonitrile ≥99%
3-Ethoxyphenylacetonitrile
Hazard: Irritant
Formula:
C₁₀H₁₁NO Boiling Pt: 103 °C (1 mmHg) Density: 1.03 |
CAS Number:
74205-55-3 |
Specification Test Results
Boiling point | 103°C/1mm |
Density | 1.03 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...