4-Ethoxy-3-methoxyphenethylamine ≥99%
Supplier: Matrix Scientific
002366-1G
101783-774EA
130.77
USD
101783-774
4-Ethoxy-3-methoxyphenethylamine ≥99%
4-Ethoxy-3-methoxyphenethylamine
Hazard: Irritant/Irritant-Harmful
Specification Test Results
Boiling point | 113°C/1mm |
Density | 1.04 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...