2-Propylpyrazine ≥97%
Supplier: Matrix Scientific
006664-5G
006664-25G
101790-878EA
60
USD
101790-878
101790-880
2-Propylpyrazine ≥97%
2-Propylpyrazine
TCA listed: T
Hazard: Irritant
Specification Test Results
Boiling point | 65°C/12mm |
Density | 0.97 |
ND20 | 1.4934 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...