1-Bromo-2,4-bis(trifluoromethyl)benzene ≥98%

Supplier: Matrix Scientific

Synonyms: 2,4-Bis(trifluoromethyl)-1-bromobenzene

005872-25G
101789-678EA 67.69 USD
101789-678
1-Bromo-2,4-bis(trifluoromethyl)benzene ≥98%
1-Bromo-2,4-bis(trifluoromethyl)benzene

Hazard: Irritant/Refrigerate

Formula: C₈H₃BrF₆
MW: 293.01 g/mol
Boiling Pt: 158 °C (740 mmHg)
Density: 1.738
MDL Number: MFCD00074904
CAS Number: 327-75-3

Order Now

Specification Test Results

Boiling point 158°C/740mm
Density 1.738
ND20 1.437

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR