4-Phenoxyphenylacetonitrile ≥98%

Supplier: Matrix Scientific

005713-5G 005713-1G
101789-412EA 78.46 USD
101789-412 101789-414
4-Phenoxyphenylacetonitrile ≥98%
4-Phenoxyphenylacetonitrile

Hazard: Irritant

Formula: C₁₄H₁₁NO
Boiling Pt: 145 °C (1 mmHg)
Density: 1.12
MDL Number: MFCD00016399
CAS Number: 92163-15-0
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 145°C/1mm
Density 1.12
ND20 1.582

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR