1-Chloro-4-(trifluoromethoxy)benzene ≥98%

Supplier: Matrix Scientific

Synonyms: 4-(Trifluoromethoxy)chlorobenzene

005348-5G
101788-712EA 34.5 USD
101788-712
1-Chloro-4-(trifluoromethoxy)benzene ≥98%
1-Chloro-4-(trifluoromethoxy)benzene

Hazard: Irritant

Formula: C₇H₄ClF₃O
MW: 196.56 g/mol
Boiling Pt: 142…145 °C
Density: 1.367
MDL Number: MFCD00040953
CAS Number: 461-81-4

Order Now

Specification Test Results

Boiling point 142-145°C
Density 1.367
ND20 1.436

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR