4-(Trifluoromethoxy)toluene ≥99%

Supplier: Matrix Scientific

005347-25G
101788-710EA 133.85 USD
101788-710
4-(Trifluoromethoxy)toluene ≥99%
4-(Trifluoromethoxy)toluene

Hazard: Flammable

Formula: C₈H₇F₃O
MW: 176.1400 g/mol
Boiling Pt: 134 °C
Density: 1.179
Flash Pt: 31 °C
MDL Number: MFCD00042413
CAS Number: 706-27-4

Order Now

Specification Test Results

Boiling point 134°C
Density 1.179
ND20 1.418

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR