2,3-Dibromo-1,1,1-trifluoropropane ≥98%

Supplier: Matrix Scientific

004526-25G 004526-5G
101787-388EA 210.77 USD
101787-388 101787-386
2,3-Dibromo-1,1,1-trifluoropropane ≥98%
2,3-Dibromo-1,1,1-trifluoropropane

TCA listed: T
Hazard: Irritant

Order Now

Specification Test Results

Boiling point 115-116°C
Density 2.117
ND20 1.429

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR