Ethyl-2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Ethoxy-2-(2,2,2-trifluoroacetyl)acrylic Acid Ethyl Ester
, Ethyl 3-Ethoxy-2-(2,2,2-trifluoroacetyl)acrylate
, Ethyl ethoxymethylene-3-oxo-4,4,4-trifluorobutyrate
, 2-(Ethoxymethylene)-4,4,4-trifluoro-3-oxobutyric Acid Ethyl Ester
008932-5G
008932-25G
101795-846EA
27.69
USD
101795-846
101795-848
Ethyl-2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate ≥97%
Ethyl-2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate
Purity: 97%
CAS: 571-55-1
MDL Number: MFCD00052716
Molecular Formula: C9H11F3O4
Molecular Weight: 240,18
Boiling Point: 80-82°C/1mm
Hazard: Irritant
Formula:
C₉H₁₁F₃O₄ MW: 240.18 g/mol Boiling Pt: 80…82 °C (1 mmHg) Density: 1.239 |
MDL Number:
MFCD02677683 CAS Number: 571-55-1 |
Specification Test Results
Boiling point | 80-82°C/1mm |
Density | 1.239 |
ND20 | 1.4262 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...