1,2-Dichloro-1,1,3,3,3-pentafluoropropane ≥97%
Supplier: Matrix Scientific
008903-5G
008903-25G
101795-774EA
200.1
USD
101795-774
101795-772
1,2-Dichloro-1,1,3,3,3-pentafluoropropane ≥97%
1,2-Dichloro-1,1,3,3,3-pentafluoropropane
Hazard: Ozone Depleter/Irritant-Harmful
Specification Test Results
Boiling point | 50.4°C |
Density | 1.5564 |
ND20 | 1.3224 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...