1H,1H,5H-Octafluoropentyl acrylate ≥97%
Supplier: Matrix Scientific
Synonyms:
2,2,3,3,4,4,5,5-Octafluoropentyl acrylate
002859-25G
101784-776EA
96.92
USD
101784-776
1H,1H,5H-Octafluoropentyl acrylate ≥97%
1H,1H,5H-Octafluoropentyl acrylate
Hazard: Irritant/Refrigerate
Formula:
C₈H₆F₈O₂ Boiling Pt: 56 °C (8 mmHg) Density: 1.481 |
MDL Number:
MFCD00039279 CAS Number: 376-84-1 |
Specification Test Results
Boiling point | 56°C/8mm |
Density | 1.481 |
ND20 | 1.346 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...