1H,1H,2H,2H-Perfluorooctyl methacrylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methacrylic acid 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-n-octyl ester
, 2-(Perfluorohexyl)ethyl methacrylate
, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-n-octyl Methacrylate
, 1H,1H,2H,2H-Tridecafluoro-n-octyl Methacrylate
, Methacrylic acid 1H,1H,2H,2H-Tridecafluoro-n-octyl ester
002242-5G
002242-25G
101783-518EA
253.85
USD
101783-518
101783-516
1H,1H,2H,2H-Perfluorooctyl methacrylate ≥95%
1H,1H,2H,2H-Perfluorooctyl methacrylate
TCA listed: T
Hazard: Irritant/Refrigerate
Formula:
C₁₂H₉F₁₃O₂ MW: 432.18 g/mol Boiling Pt: 92 °C (8 mmHg) Density: 1.496 |
MDL Number:
MFCD00077580 CAS Number: 2144-53-8 |
Specification Test Results
Boiling point | 92°C/8mm |
Density | 1.496 |
ND20 | 1.346 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...