4-(1,1,2,2-Tetrafluoroethoxy)toluene ≥98%

Supplier: Matrix Scientific

002038-25G 002038-5G
101783-086EA 136 USD
101783-086 101783-084
4-(1,1,2,2-Tetrafluoroethoxy)toluene ≥98%
4-(1,1,2,2-Tetrafluoroethoxy)toluene

Purity: 98%
CAS: 1737-11-7
MDL Number: MFCD00042105
Molecular Formula: C9H8F4O
Molecular Weight: 208,16
Boiling Point: 67-69°C/14mm
Hazard: Irritant

Order Now

Specification Test Results

Boiling point 67-69°C/14m
Density 1.25
ND20 1.422

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR