4-(1,1,2,2-Tetrafluoroethoxy)aniline ≥98%
Supplier: Matrix Scientific
002033-1G
101783-082EA
75.38
USD
101783-082
4-(1,1,2,2-Tetrafluoroethoxy)aniline ≥98%
4-(1,1,2,2-Tetrafluoroethoxy)aniline
Hazard: Irritant
Specification Test Results
Boiling point | 104-108°C/1 |
Density | 1.379 |
ND20 | 1.462 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...