1-Chloro-2,3,5,6-tetrafluorobenzene ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Chloro-1,2,4,5-tetrafluorobenzene
001939-25G
101782-890EA
444.62
USD
101782-890
1-Chloro-2,3,5,6-tetrafluorobenzene ≥97%
1-Chloro-2,3,5,6-tetrafluorobenzene
Hazard: Irritant
Specification Test Results
Boiling point | 125-126°C/7 |
Density | 1,538 |
ND20 | 1.442 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...