1,1,2,3,3,3-Hexafluoropropyl ethyl ether ≥97%
Supplier: Matrix Scientific
Synonyms:
Ethyl 1,1,2,3,3,3-Hexafluoropropyl ether
001195-5G
101781-122EA
121.54
USD
101781-122
1,1,2,3,3,3-Hexafluoropropyl ethyl ether ≥97%
1,1,2,3,3,3-Hexafluoropropyl ethyl ether
Hazard: Irritant
Specification Test Results
Boiling point | 71°C |
Density | 1.321 |
ND20 | 1.305 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...