9-Bromononanoic acid ≥98%

Supplier: Matrix Scientific

007524-1G
101792-982EA 75.85 USD
101792-982
9-Bromononanoic acid ≥98%
9-Bromononanoic acid

Purity: 98%
CAS: 41059-02-3
MDL Number: MFCD00046564
Molecular Formula: C9H17BrO2
Molecular Weight: 237,14
Boiling Point: 165°C/3mm
Hazard: Corrosive

Formula: C₉H₁₇BrO₂
MW: 237,14 g/mol
Boiling Pt: 165 °C (3 mmHg)
Melting Pt: 39 °C
MDL Number: MFCD00046564
CAS Number: 41059-02-3

Order Now

Specification Test Results

Boiling point 165°C/3mm
Melting point 39°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR