3-(Trifluoromethyl)benzhydrol ≥97%

Supplier: Matrix Scientific

Synonyms: Phenyl(3-(trifluoromethyl)phenyl)methanol

005461-1G
101788-992EA 16.1 USD
101788-992
3-(Trifluoromethyl)benzhydrol ≥97%
3-(Trifluoromethyl)benzhydrol

Hazard: Irritant

Formula: C₁₄H₁₁F₃O
MW: 252.23 g/mol
Boiling Pt: 125…127 °C (0.5 mmHg)
Melting Pt: 63…64 °C
MDL Number: MFCD00014397
CAS Number: 728-80-3

Order Now

Specification Test Results

Boiling point 125-127°C/0
Melting point 63-64°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR