1-(Pentafluorophenyl)ethanol ≥97%
Supplier: Matrix Scientific
Synonyms:
(1-Hydroxyethyl)pentafluorobenzene
004425-25G
004425-5G
101787-222EA
89.23
USD
101787-222
101787-220
1-(Pentafluorophenyl)ethanol ≥97%
1-(Pentafluorophenyl)ethanol
Hazard: Irritant
Formula:
C₈H₅F₅O MW: 212.12 g/mol Boiling Pt: 102…103 °C (30 mmHg) Melting Pt: 34…37 °C |
MDL Number:
MFCD00004510 CAS Number: 830-50-2 |
Specification Test Results
Boiling point | 102-103°C/3 |
Melting point | 34-37°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...