Methyl-3-aminothiophene-2-carboxylate ≥98%

Supplier: Matrix Scientific

Synonyms: Methyl 3-amino-2-thiophenecarboxylate

018289-100G
101805-842EA 39.1 USD
101805-842
Methyl-3-aminothiophene-2-carboxylate ≥98%
Methyl 3-aminothiophene-2-carboxylate

Hazard: Irritant

Formula: C₆H₇NO₂S
MW: 157.19 g/mol
Boiling Pt: 100…102 °C (0.1 mmHg)
Melting Pt: 62…64 °C
MDL Number: MFCD00009765
CAS Number: 22288-78-4

Order Now

Specification Test Results

Boiling point 100-102°C/0
Melting point 62-64°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR