3'-Isothiocyanatoacetophenone ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Acetylphenyl isothiocyanate

003877-5G
101786-356EA 366.85 USD
101786-356
3'-Isothiocyanatoacetophenone ≥98%
3'-Isothiocyanatoacetophenone

Hazard: Toxic/Corrosive/Moisture Sensitive

Formula: C₉H₇NOS
Boiling Pt: 135…136 °C (2 mmHg)
Melting Pt: 24 °C
MDL Number: MFCD00041081
CAS Number: 3125-71-1

Order Now

Specification Test Results

Boiling point 135-136°C/2
Melting point 26-28
ND20 1.648

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR